Skip to Content
Merck

BDPH9C021A80

Bide Pharmatech

6,7,8,9-Tetrahydro-5H-benzo[7]annulene-7-carboxylic acid

Synonym(s):

6,7,8,9-tetrahydro-5H-benzo[7]annulene-7-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H14O2
Molecular Weight:
190.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H14O2/c13-12(14)11-7-5-9-3-1-2-4-10(9)6-8-11/h1-4,11H,5-8H2,(H,13,14)

Inchi Key

FGVCREOLYLRJCU-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library