Merck
  • Determination of glucose in plasma by dry film-based Fourier transformed-infrared spectroscopy coupled with boosting support vector regression.
Determination of glucose in plasma by dry film-based Fourier transformed-infrared spectroscopy coupled with boosting support vector regression.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2007-07-13)
Yan-Ping Zhou, Lu Xu, Li-Juan Tang, Jian-Hui Jiang, Guo-Li Shen, Ru-Qin Yu, Yukihiro Ozaki
ABSTRACT
In the present study, a dry film-based Fourier transformed-infrared (FT-IR) spectroscopic technique, coupled with boosting support vector regression (BSVR), was employed for a blood glucose assay. Potassium thiocyanate (KSCN) was taken in the dry-film method as an internal standard to compensate for any film thickness variation. This technique circumvents interference from water absorption, and requires only 5 microl of a sample. Moving window partial least-squares regression (MWPLSR) was used for wavenumber interval selection before multivariate modeling. By using the BSVR modeling technique, glucose in plasma could be determined over a 0.4 - 20 mmol/l concentration range with satisfactory accuracy. The performance of the BSVR methodology was compared with that of conventional support vector regression (SVR) as well as partial-least squares (PLS). The results demonstrated that BSVR is an effective multivariate calibration tool, providing better performance than conventional PLS and SVR.
Sigma-Aldrich
Potassium thiocyanate solution, volumetric, 8 M KSCN
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium thiocyanate, ACS reagent, 99%
Sigma-Aldrich
Potassium thiocyanate, ReagentPlus®, ≥99.0%