Merck
Deciphering the proteomic signature of human endometrial receptivity.
Human reproduction (Oxford, England) (2014-08-12)
Tamara Garrido-Gómez, Alicia Quiñonero, Oreto Antúnez, Patricia Díaz-Gimeno, Jose Bellver, Carlos Simón, Francisco Domínguez
ABSTRACT
Are there any proteomic differences between receptive (R) and non-receptive (NR) endometrial receptivity array (ERA)-diagnosed endometria obtained on the same day of a hormonal replacement therapy (HRT) treatment cycle? There is a different proteomic signature between R and NR ERA-diagnosed endometrium obtained on the same day of HRT cycles. The human endometrial transcriptome has been extensively investigated in the last decade resulting in the development of a new diagnostic test based on the transcriptomic signature of the window of implantation (WOI). Much less is known about the proteomics derived from the transcripts present during the WOI. This study was a basic proteomic analysis of human endometrial biopsies taken from twelve IVF patients. Human endometrial biopsies were collected during HRT cycles after 5 days of progesterone (P) administration, and diagnosed as receptive (R; n = 6) or non-receptive (NR; n = 6) by the ERA test. Endometrial proteins were extracted, labelled and separated using differential in-gel electrophoresis (DIGE). Proteins were identified using mass spectrometry, followed up by in silico analysis. Validation studies using western blots and immunolocalization were performed for the progesterone receptor membrane component 1 (PGRMC1) and annexin A6 (ANXA6) proteins. DIGE analysis followed by protein identification by MALDI-MS and database searches revealed 24 differentially expressed proteins in R versus NR samples. In silico analysis showed two pathways which were significantly different between R and NR samples. Expression of PGRMC1 and ANXA6 was validated and localized by western blots and immunohistochemistry. These results highlight these proteins as key targets likely to be important in the comprehension of human endometrial receptivity. This was mainly a descriptive study with no functional studies on the proteins found. We also used a low number of human endometrial samples for the DIGE analysis. This study identified the proteomic profile associated with receptive or non-receptive human endometria. Our findings suggest that although histological dating indicates a putative 'receptive' status within the WOI, a different transcriptomic and proteomic profile is observed in these samples. We should move towards using more personalized WOIs, where identification of the correct endometrial receptivity status, and consequently the success of IVF, relies on individual molecular signatures rather than traditional endometrial dating. F.D.'s participation in this work was supported by the Spanish Ministry of Economy and Competitiveness, through the Miguel Servet Programme (CP13/00075) co-founded by FEDER. The project was also supported by a grant from the Spanish Ministry of Economy and Competitiveness, through the FIS Programme (PI12/00450). The authors have no financial/commercial conflicts of interest to declare.
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
SAFC
Glycine
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.