Merck
  • X-ray Spectroscopy Uncovering the Effects of Cu Based Nanoparticle Concentration and Structure on Phaseolus vulgaris Germination and Seedling Development.
X-ray Spectroscopy Uncovering the Effects of Cu Based Nanoparticle Concentration and Structure on Phaseolus vulgaris Germination and Seedling Development.
Journal of agricultural and food chemistry (2017-08-18)
Nádia M Duran, Susilaine M Savassa, Rafael Giovanini de Lima, Eduardo de Almeida, Francisco S Linhares, Cornelis A M van Gestel, Hudson W Pereira de Carvalho
ABSTRACT
Nanoparticles properties such as solubility, tunable surface charges, and singular reactivity might be explored to improve the performance of fertilizers. Nevertheless, these unique properties may also bring risks to the environment since the fate of nanoparticles is poorly understood. This study investigated the impact of a range of CuO nanoparticles sizes and concentrations on the germination and seedling development of Phaseolus vulgaris L. Nanoparticles did not affect seed germination, but seedling weight gain was promoted by 100 mg Cu L