MilliporeSigma
Faster. Simpler. More
Efficient Purification.
Flow-through polishing for higher yields and streamlined workflows.
HomeStreamline Purification with Flow Through Polishing Platforms
Streamline Purification with Flow Through Polishing Platforms
What if you could simplify your purification process, boost efficiency, and maximize yield - all while cutting costs? By integrating flow-through polishing resin or membrane technologies into an intensified workflow, you can overcome the limitations of bind-elute chromatography and take your process to the next level.
Multiple containers of Eshmuno® CP-FT cation exchange resin for efficient mAb aggregate removal in flow-through chromatography.
Eshmuno® CP-FT cation exchange (CEX) resin enables highly efficient removal of mAb aggregates in flow-through frontal chromatography mode. With loading capacities up to 10× higher than traditional bind/elute CEX chromatography, it supports higher throughput and greater process efficiency, making it an ideal contributor to process intensification. Learn more.
Different sizes of Natrix® CH chromatography membrane devices for high-capacity aggregate removal in frontal chromatography.
Natrix® CH chromatography membrane overcomes the limitations of bind-elute purification by efficiently removing aggregates in frontal chromatography mode. Its exceptional loading capacity of up to 1000 g/L at very high flow rates result in an unprecedented productivity. Natrix® CH is a ready-to-use device you can use to accelerate, intensify and simplify your purification process. Learn more.
Related Resources
Try it for yourself – experience the benefits of process intensification!
Fill out the form below to request a Natrix® CH device or Eshmuno® CP-FT resin sample. Fields with an * are required.
Flow-Through Polishing Trial Offer Official Rules