Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

BDPH9BCDFDCD

Bide Pharmatech

4-Chloro-6,8-difluoro-2-methylquinoline

Synonym(s):

4-chloro-6,8-difluoro-2-methylquinoline

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFDCD is not currently sold in your country.

About This Item

Linear Formula:
C10H6N1Cl1F2
CAS Number:
Molecular Weight:
213.61
MDL number:
Product BDPH9BCDFDCD is not currently sold in your country.

Inchi Code

1S/C10H6ClF2N/c1-5-2-8(11)7-3-6(12)4-9(13)10(7)14-5/h2-4H,1H3

Inchi Key

AFFPXAYTKXVJTD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals or flocculent solid

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Danger

pictograms

CorrosionExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library