MilliporeSigma
Synchronization and superovulation of mature cycling gilts for the collection of pronuclear stage embryos.
Animal reproduction science (2006-11-23)
Jeffrey R Sommer, E Bruce Collins, Jose L Estrada, Robert M Petters
RÉSUMÉ
An efficient protocol was developed to synchronize and superovulate mature pigs for the collection of pronuclear stage embryos suitable for DNA microinjection. A timed and coordinated regimen of Lutalyse, PG600 and Chorulon along with daily checking for estrus allowed synchronization of groups of gilts having estrous cycles at regular intervals. Pigs 10-16 days after the beginning of standing estrus have been successfully synchronized into estrus using this protocol. A standard dose of each drug was used independent of size or age of the animal. One protocol averaged 38.9 ovulations and 31.1 one-cell embryos recovered per animal.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Prostaglandin F tris salt, ≥99%, synthetic, powder
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Sigma-Aldrich
Prostaglandin F tris salt, synthetic, suitable for cell culture
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Dinoprost trometamol, European Pharmacopoeia (EP) Reference Standard
Le tarif et la disponibilité ne sont pas disponibles actuellement.