MilliporeSigma
Cumulative sensitization and disease in a beryllium oxide ceramics worker cohort.
Journal of occupational and environmental medicine (2008-12-19)
Christine R Schuler, Margaret M Kitt, Paul K Henneberger, David C Deubner, Kathleen Kreiss
RÉSUMÉ
We followed a cohort of 136 beryllium oxide ceramics workers from 1992 to 2003, including those who left employment, for beryllium sensitization and chronic beryllium disease (CBD). We invited the cohort's participation in current worker surveys in 1992, 1998, 2000, and 2002-2003, and in former worker surveys in 2000-2001 and 2003. We calculated 11-year cumulative incidences (after 1992 initial survey) of sensitization and CBD, both crude and corrected for interval censoring; and period prevalences (including 1992 findings), crude and corrected. In 1992, point prevalences were 6% sensitized and 4% CBD. We obtained follow-up on 83% of 128 not sensitized in 1992. Crude cumulative incidences for sensitization and CBD were 13% and 9%, respectively; corrected were 15% and 11%. Crude period prevalences for sensitization and CBD were 16% and 11%, respectively; corrected were 20% and 14%. Corrected period prevalences for pre-1992 machining work were 30% and 20%. With repeated testing over 11 years, total sensitization and CBD in this cohort were triple initial 1992 survey results.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Beryllium oxide, 99.98% trace metals basis
Le tarif et la disponibilité ne sont pas disponibles actuellement.