MilliporeSigma
Expression and function of GSTA1 in lung cancer cells.
Asian Pacific journal of cancer prevention : APJCP (2014-11-07)
Xue-Diao Pan, Zhou-Ping Yang, Qi-Ling Tang, Tong Peng, Zheng-Bing Zhang, Si-Gui Zhou, Gui-Xiang Wang, Bing He, Lin-Quan Zang
RÉSUMÉ
Glutathione S-transferase A1 (GSTA1) appears to be primarily involved in detoxification processes, but possible roles in lung cancer remain unclear. The objective of this study was to investigate the expression and function of GSTA1 in lung cancer cells. Real-time PCR and Western blotting were performed to assess expression in cancer cell lines and the normal lung cells, then verify the A549 cells line with stable overexpression. Localization of GSTA1 proteins was assessed by cytoimmunofluorescence. Three double-strand DNA oligoRNAs (SiRNAs) were synthesized prior to being transfected into A549 cells with Lipofectamine 2000, and then the most efficient SiRNA was selected. Expression of the GSTA1 gene in the transfected cells was determined by real-time PCR and Western blotting. The viability of the transfected cells were assessed by MTT. Results showed that the mRNA and protein expression of A549 cancer cells was higher than in MRC-5 normal cells. Cytoimmunofluorescence demonstrated GSTA1 localization in the cell cytoplasm and/or membranes. Transfection into A549 cells demonstrated that down-regulated expression could inhibit cell viability. Our data indicated that GSTA1 expression may be a target molecule in early diagnosis and treatment of lung cancer.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
GST A1-1, Recombinant Human
Le tarif et la disponibilité ne sont pas disponibles actuellement.