MilliporeSigma
  • The potassium channel KCa3.1 constitutes a pharmacological target for neuroinflammation associated with ischemia/reperfusion stroke.
The potassium channel KCa3.1 constitutes a pharmacological target for neuroinflammation associated with ischemia/reperfusion stroke.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2015-12-15)
Yi-Je Chen, Hai M Nguyen, Izumi Maezawa, Eva M Grössinger, April L Garing, Ralf Köhler, Lee-Way Jin, Heike Wulff
RÉSUMÉ
Activated microglia/macrophages significantly contribute to the secondary inflammatory damage in ischemic stroke. Cultured neonatal microglia express the K
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Anti-KCNN4 antibody produced in rabbit, affinity isolated antibody
Le tarif et la disponibilité ne sont pas disponibles actuellement.