Skip to Content
Merck

GF62817824

Aluminum

foil, thickness 0.020 mm, diameter 8 mm, purity 99%

Synonym(s):

AL000405

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

99%

form

foil

autoignition temp.

1400 °F

manufacturer/tradename

Goodfellow 628-178-24

resistivity

2.6548 μΩ-cm

diam.

8 mm

thickness

0.020 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF72541379GF68640363GF84319487
assay

99%

assay

99%

assay

99%

assay

99%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 628-178-24

manufacturer/tradename

Goodfellow 725-413-79

manufacturer/tradename

Goodfellow 686-403-63

manufacturer/tradename

Goodfellow 843-194-87

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service