Skip to Content

BDPH9BCD4E5D

Bide Pharmatech

(4-Methylnaphthalen-1-yl)methanol

Synonym(s):

(4-methyl-1-naphthyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H12O1
Molecular Weight:
172.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H12O/c1-9-6-7-10(8-13)12-5-3-2-4-11(9)12/h2-7,13H,8H2,1H3

Inchi Key

RRSGUDDGNKMFRY-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library