Skip to Content
Merck

BDPH9BCD6959

Bide Pharmatech

4'-(1-(2-Benzylpiperidine-1-carbonyl)-1H-1,2,3-triazol-4-yl)-[1,1'-biphenyl]-3-carboxylic acid

Synonym(s):

4'-(1-(2-benzylpiperidine-1-carbonyl)-1H-1,2,3-triazol-4-yl)-[1,1'-biphenyl]-3-carboxylic acid

Select a Size

Change View

About This Item

Linear Formula:
C28H26O3N4
CAS Number:
Molecular Weight:
466.54
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C28H26N4O3/c33-27(34)24-10-6-9-23(18-24)21-12-14-22(15-13-21)26-19-32(30-29-26)28(35)31-16-5-4-11-25(31)17-20-7-2-1-3-8-20/h1-3,6-10,12-15,18-19,25H,4-5,11,16-17H2,(H,33,34)

Inchi Key

SSSCOJOXPDDHOO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid






Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library