Skip to Content
Merck

BDPH9BCD7828

Bide Pharmatech

6-(Dimethoxymethyl)nicotinaldehyde

Synonym(s):

6-(dimethoxymethyl)nicotinaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O3N1
CAS Number:
Molecular Weight:
181.19
MDL number:
Product BDPH9BCD7828 is not currently sold in your country.


Inchi Code

1S/C9H11NO3/c1-12-9(13-2)8-4-3-7(6-11)5-10-8/h3-6,9H,1-2H3

Inchi Key

ZZGVXCUKGWMGOM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or semi-solid or solid



Still not finding the right product?

Explore all of our products under 6-(Dimethoxymethyl)nicotinaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library