Skip to Content

BDPH9BCDCD2F

Bide Pharmatech

Methyl 5-amino-2-methoxybenzoate

Synonym(s):

methyl 5-amino-2-methoxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O3N1
CAS Number:
Molecular Weight:
181.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO3/c1-12-8-4-3-6(10)5-7(8)9(11)13-2/h3-5H,10H2,1-2H3

Inchi Key

PSCXCIPPRCFAAO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?

Explore all of our products under Methyl 5-amino-2-methoxybenzoate


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library