Skip to Content

BDPH9C021BD4

Bide Pharmatech

(4-(Trifluoromethoxy)phenyl)methanethiol

Synonym(s):

4-(trifluoromethoxy)benzyl hydrosulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H7O1S1F3
Molecular Weight:
208.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H7F3OS/c9-8(10,11)12-7-3-1-6(5-13)2-4-7/h1-4,13H,5H2

Inchi Key

VWLQXXSOXQNHJM-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library