Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H13NO.ClH/c1-7(11)9(10)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9+;/m0./s1
Inchi Key
VIOJYFJDMKDXOT-DKXTVVGFSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
White to yellow powder or crystals
Still not finding the right product?
Explore all of our products under (1S,2S)-1-Amino-1-phenylpropan-2-ol hydrochloride
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
