Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
UP-158-22-3-10, monoclonal
species reactivity
rat, human, mouse
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... MAPK3(5595)
General description
Mitogen-activated protein kinase 3 (EC 2.7.11.24; UniProt P21708; also known as ERK-1, ERT2, Extracellular signal-regulated kinase 1, Insulin-stimulated MAP2 kinase, MAP kinase 3, MAP kinase isoform p44, MAPK 3, Microtubule-associated protein 2 kinase, MNK1, p44-ERK1, p44-MAPK) is encoded by the Mapk3 (also known as Erk1, Prkm3) gene (Gene ID 50689) in rat species. ERK1 (a.k.a. MAPK 3) and ERK2 (a.k.a. MAPK 1 or MAPK 2) are related serine/threonine kinases of the Ras-Raf-MEK-ERK signaling pathway. ERK1/2 are activated by MEK1/2-catalyzed phosphorylation at their conserved Thr-Glu-Tyr (TEY) dual phosphorylation motif, first at the Tyr residue (human/rat ERK1 Tyr204/Tyr203, human/rat ERK2 Tyr187/Tyr183) and then at the Thr residue (human/rat ERK1 Thr202/Thr201, human/rat ERK2 Thr185/Thr181). ERK1/2 are proline-directed kinases that preferentially catalyze the phosphorylation of Pro-Xxx-Ser/Thr-Pro sequence motif in hundreds of cytoplasmic and nuclear substrates, including transcription factors such as Ets, Elk, and c-Fos. Besides this primary structure requirement, many ERK1/2 substrates possess a D-docking site and/or an F-docking site. A variety of scaffold proteins, including KSR1/2, IQGAP1, MP1, and -Arrestin1/2, also participate in the regulation of ERK1/2 MAP kinase cascade. Ras-Raf-MEK-ERK signaling activity is upregulated in about one-third of all human cancers, and targeted inhibition against components of this signaling pathway represents a popular anticancer strategy.
~44 kDa observed. 43.00/38.14 kDa (human MAPK3 isoform 1/2), 42.95/45.64 kDa (rat MAPK3 isoform 1/2), 42.93 kDa (mouse MAPK3) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).
Immunogen
KLH-conjugated linear peptide corresponding to the C-terminal sequence of rat ERK-1 (MAPK 3).
Application
Immunoprecipitation Analysis: A representative lot immunoprecipitated active Erk1 from lysate of NGF-stimulated (50 ng/mL) PC-12 rat pheochromocytoma cells.
Western Blotting Analysis: A representative lot detected Erk1 in human MCF-7 is a breast cancer cell lysate (Chen, J.Q., et al. (2013). Anal. Biochem. 442(1):97-103).
Western Blotting Analysis: A representative lot detected Erk1 in human HaCat keratinocyte lysate (Jarosz, M., et al. (2012). PLoS One. 7(6):e39436).
Western Blotting Analysis: A representative lot detected Erk1 in HT-29 human colorectal adenocarcinoma cell lysate (O′Neill, R.A., et al. (2006). Proc. Natl. Acad. Sci. U. S. A. 103(44):16153-16158).
Western Blotting Analysis: A representative lot detected Erk1 in human MCF-7 is a breast cancer cell lysate (Chen, J.Q., et al. (2013). Anal. Biochem. 442(1):97-103).
Western Blotting Analysis: A representative lot detected Erk1 in human HaCat keratinocyte lysate (Jarosz, M., et al. (2012). PLoS One. 7(6):e39436).
Western Blotting Analysis: A representative lot detected Erk1 in HT-29 human colorectal adenocarcinoma cell lysate (O′Neill, R.A., et al. (2006). Proc. Natl. Acad. Sci. U. S. A. 103(44):16153-16158).
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
This rabbit monoclonal Anti-ERK1/MAPK3 Antibody, CT, clone UP-158-22-3-10, Cat. No. 05-957-I detects levels of ERK1/MAPK3, and has been published and validated for use in Immunoprecipitation and Western Blotting.
Biochem/physiol Actions
Clone UP-158-22-3-10 recognizes a C-terminal epitope of MAPK 3 (ERK1). Immunogen sequence is not present in human MAPK3 spliced isoform 3 (ERK1b).
Physical form
Rabbit monoclonal antibody IgG hybridoma culture supernatant without preservatives.
Unpurified.
Preparation Note
Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Evaluated by Western Blotting in NIH/3T3 cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this hybridoma culture supernatant detected Erk1 in 10 µg of mouse NIH/3T3 cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this hybridoma culture supernatant detected Erk1 in 10 µg of mouse NIH/3T3 cell lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service