About This Item
Skip To
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
7H8.1, monoclonal
species reactivity
human
technique(s)
western blot: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... DBF4(10926)
1 of 4
This Item | SAB4500178 | SAB1403376 | SAB2501760 |
|---|---|---|---|
| species reactivity human | species reactivity human, mouse | species reactivity human | species reactivity rat, mouse |
| antibody form purified antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| Gene Information human ... DBF4(10926) | Gene Information human ... DBF4(10926) | Gene Information human ... DBF4B(80174) | Gene Information mouse ... Dbf4(27214) |
| clone 7H8.1, monoclonal | clone polyclonal | clone 1A7, monoclonal | clone polyclonal |
| biological source mouse | biological source rabbit | biological source mouse | biological source goat |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
General description
Immunogen
Application
Epigenetics & Nuclear Function
Chromatin Biology
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 0.5 µg/mL of this antibody detected DBF4 in 200 µg of HepG2 cell lysate.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


