Product Name
Natrix® Process Holder,
sterility
non-sterile
Quality Level
product line
Natrix®
matrix
Stainless-steel base material
application(s)
pharmaceutical
separation technique
cation exchange
Related Categories
General description
The Natrix® Process holder is an equipment where up to 10 Pilot and/or Process devices can be stacked together.
Legal Information
NATRIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service