Skip to Content
Merck

STS0028

ECOSURF EH-9 solution

greener alternative

90% ( aqueous solution), suitable for molecular biology

Synonym(s):

2-Ethyl hexanol EoPo copolymer (9 EO), Alcohol (2EH) alkoxylate, Ethyl hexanol ethoxylated propoxylated

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

90% ( aqueous solution)

CMC

1066 ppm (25°C)

viscosity

61.8 cSt(40 °C)

transition temp

pour point -5 °C
cloud point 64 °C (10 wt% actives aq solution)

solubility

water: soluble

density

1.0157 g/mL at 40 °C

HLB

12.5

suitability

suitable for molecular biology

application(s)

life science and biopharma

foreign activity

Dnase, Rnase, none detected

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

This product is a 90 % aqueous solution of an alcohol ethoxylate

Features and Benefits

  • Superior wetting
  • Excellent oily soil removal
  • Rapid dissolution & good rinseability
  • Low odor
  • No gel range
  • Outstanding formulation & handling properties
  • Readily biodegradable
  • Aquatic toxicity EC50 > 10 mg/L

Legal Information

ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow
ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service