Product Name
Corning® Falcon® Cell Culture Dish, plate diam. × capacity 35 x 10 mm × 2.5 mL, Tissue Culture (TC)-treated surface, sterile
sterility
sterile
packaging
case of 500 ea (25 packs of 20)
manufacturer/tradename
Corning 353001
plate diam. × capacity
35 x 10 mm × 2.5 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Cell Culture
General description
Falcon® 35mm x 10mm Style Easy-Grip Cell Culture Dish, Tissue Culture-Treated, Sterile, Bulk Packaged, 20/Pack, 500/Case
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service