Skip to Content
Merck

SAB2105303

Anti-FOXA2, (C-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HNF3-beta, Anti-HNF3beta, Anti-Hnf-3b, Anti-Hnf3b, Anti-Tcf-3b

Sign In to View Organizational & Contract Pricing

Select a Size

100 μL

CHF 419.00

CHF 419.00


Check Cart for Availability


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXA2(3170)
mouse ... Foxa2(15376)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
AB4125SAB2100835HPA066846
conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

species reactivity

mouse, human

species reactivity

rat, human, mouse

species reactivity

bovine, mouse, human, rat, dog, pig

species reactivity

human

Immunogen

Synthetic peptide directed towards the C terminal of human Foxa2

Biochem/physiol Actions

Foxa2 is a transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression and regulation of gene expression in differentiated tissues. Foxa2 is thought to act as a ′pioneer′ factor opening the compacted chromatin for other proteins through interactions with nucleosomal core histones and thereby replacing linker histones at target enhancer and/or promoter sites. In embryonic development Foxa2 is required for notochord formation. Foxa2 is involved in the development of multiple endoderm-derived organ systems such as the liver, pancreas and lungs; Foxa1 and Foxa2 seem to have at least in part redundant roles. Foxa2 modulates the transcriptional activity of nuclear hormone receptors; inhibits AR-mediated transcription from the LCN5 promoter. Foxa2 binds to fibrinogen beta promoter and is involved in IL6-induced fibrinogen beta transcriptional activation. Foxa2 interacts with the cis-acting regulatory regions of these genes. Foxa2 is involved in glucose homeostasis; regulates the expression of genes important for glucose sensing in pancreatic beta-cells and glucose homeostasis. Foxa2 is involved in regulation of fat metabolism; activates transcriptional programs of lipid metabolism and ketogenesis at low insulin state.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: VDVEGTDYLNGDLGWSSSVSDSDERGSMQSLGSDEGYSSATVKRAKLQDG

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service