Skip to Content
Merck

V520829

Palladium on carbon

extent of labeling: 10 wt. % loading (dry basis), matrix carbon powder, solid, wet support, Vetec

Synonym(s):

Pd/C

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
Pd
Molecular Weight:
106.42
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:
NACRES:
NA.22
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Palladium on carbon, extent of labeling: 10 wt. % loading (dry basis), matrix carbon powder, wet support, Vetec

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

form

solid
wet

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

extent of labeling

10 wt. % loading (dry basis)

matrix

carbon powder, wet support

Quality Level

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenation of alkenes, alkynes, ketones, nitriles, imines, azides, nitro groups, benzenoid and heterocyclic aromatics; used for hydrogenolysis of cyclopropanes, benzyl derivatives, epoxides, hydrazines, and halides; used to dehydrogenate aromatics and deformylate aldehydes.
Catalyst for:
  • Stille reaction
  • Hydrogenation reactions
  • Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions
  • Deoxygenation
  • Oxidation reactions
  • Coupling reactions

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishad Thamban Chandrika et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-28)
The fact that the number of people with Alzheimer's disease is increasing, combined with the limited availability of drugs for its treatment, emphasize the need for the development of novel effective therapeutics for treating this brain disorder. Herein, we focus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service