Merck
Expansion Microscopy for Cell Biology Analysis in Fungi.
Frontiers in microbiology (2020-04-23)
Ralph Götz, Sabine Panzer, Nora Trinks, Janna Eilts, Johannes Wagener, David Turrà, Antonio Di Pietro, Markus Sauer, Ulrich Terpitz
ABSTRACT
Super-resolution microscopy has evolved as a powerful method for subdiffraction-resolution fluorescence imaging of cells and cellular organelles, but requires sophisticated and expensive installations. Expansion microscopy (ExM), which is based on the physical expansion of the cellular structure of interest, provides a cheap alternative to bypass the diffraction limit and enable super-resolution imaging on a conventional fluorescence microscope. While ExM has shown impressive results for the magnified visualization of proteins and RNAs in cells and tissues, it has not yet been applied in fungi, mainly due to their complex cell wall. Here we developed a method that enables reliable isotropic expansion of ascomycetes and basidiomycetes upon treatment with cell wall degrading enzymes. Confocal laser scanning microscopy (CLSM) and structured illumination microscopy (SIM) images of 4.5-fold expanded sporidia of Ustilago maydis expressing fluorescent fungal rhodopsins and hyphae of Fusarium oxysporum or Aspergillus fumigatus expressing either histone H1-mCherry together with Lifeact-sGFP or mRFP targeted to mitochondria, revealed details of subcellular structures with an estimated spatial resolution of around 30 nm. ExM is thus well suited for cell biology studies in fungi on conventional fluorescence microscopes.
Sigma-Aldrich
Ethylenediaminetetraacetic acid dipotassium salt dihydrate, ≥98%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Anti-Rabbit-IgG - Atto 647N antibody produced in goat, 1 mg/mL IgG
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Millipore
Potato Dextrose Broth, NutriSelect® Basic, powder, for yeasts, for bacteria, for molds, pack of 250 g, pkg of 1 kg
Sigma-Aldrich
Sodium acrylate, 97%
Sigma-Aldrich
Acrylamide/Bis-acrylamide, 40% solution, BioReagent, suitable for electrophoresis, 19:1