Merck
  • Structural characterization of KKT4, an unconventional microtubule-binding kinetochore protein.
Structural characterization of KKT4, an unconventional microtubule-binding kinetochore protein.
Structure (London, England : 1993) (2021-04-30)
Patryk Ludzia, Edward D Lowe, Gabriele Marcianò, Shabaz Mohammed, Christina Redfield, Bungo Akiyoshi
ABSTRACT
The kinetochore is the macromolecular machinery that drives chromosome segregation by interacting with spindle microtubules. Kinetoplastids (such as Trypanosoma brucei), a group of evolutionarily divergent eukaryotes, have a unique set of kinetochore proteins that lack any significant homology to canonical kinetochore components. To date, KKT4 is the only kinetoplastid kinetochore protein that is known to bind microtubules. Here we use X-ray crystallography, NMR spectroscopy, and crosslinking mass spectrometry to characterize the structure and dynamics of KKT4. We show that its microtubule-binding domain consists of a coiled-coil structure followed by a positively charged disordered tail. The structure of the C-terminal BRCT domain of KKT4 reveals that it is likely a phosphorylation-dependent protein-protein interaction domain. The BRCT domain interacts with the N-terminal region of the KKT4 microtubule-binding domain and with a phosphopeptide derived from KKT8. Taken together, these results provide structural insights into the unconventional kinetoplastid kinetochore protein KKT4.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
EDTA, Disodium Salt, Dihydrate, Molecular Biology Grade
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%