Merck
  • Immunosuppressive Myeloid Cells Induce Nitric Oxide-Dependent DNA Damage and p53 Pathway Activation in CD8+ T Cells.
Immunosuppressive Myeloid Cells Induce Nitric Oxide-Dependent DNA Damage and p53 Pathway Activation in CD8+ T Cells.
Cancer immunology research (2021-01-31)
Adam N R Cartwright, Shengbao Suo, Soumya Badrinath, Sushil Kumar, Johannes Melms, Adrienne Luoma, Archis Bagati, Assieh Saadatpour, Benjamin Izar, Guo-Cheng Yuan, Kai W Wucherpfennig
ABSTRACT
Tumor-infiltrating myeloid-derived suppressor cells (MDSC) are associated with poor survival outcomes in many human cancers. MDSCs inhibit T cell-mediated tumor immunity in part because they strongly inhibit T-cell function. However, whether MDSCs inhibit early or later steps of T-cell activation is not well established. Here we show that MDSCs inhibited proliferation and induced apoptosis of CD8+ T cells even in the presence of dendritic cells (DC) presenting a high-affinity cognate peptide. This inhibitory effect was also observed with delayed addition of MDSCs to cocultures, consistent with functional data showing that T cells expressed multiple early activation markers even in the presence of MDSCs. Single-cell RNA-sequencing analysis of CD8+ T cells demonstrated a p53 transcriptional signature in CD8+ T cells cocultured with MDSCs and DCs. Confocal microscopy showed induction of DNA damage and nuclear accumulation of activated p53 protein in a substantial fraction of these T cells. DNA damage in T cells was dependent on the iNOS enzyme and subsequent nitric oxide release by MDSCs. Small molecule-mediated inhibition of iNOS or inactivation of the Nos2 gene in MDSCs markedly diminished DNA damage in CD8+ T cells. DNA damage in CD8+ T cells was also observed in KPC pancreatic tumors but was reduced in tumors implanted into Nos2-deficient mice compared with wild-type mice. These data demonstrate that MDSCs do not block early steps of T-cell activation but rather induce DNA damage and p53 pathway activation in CD8+ T cells through an iNOS-dependent pathway.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
NG-Methyl-L-arginine acetate salt, ≥98% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Roche
DNase I, from bovine pancreas