Merck
  • Decoding the molecular crosstalk between grafted stem cells and the stroke-injured brain.
Decoding the molecular crosstalk between grafted stem cells and the stroke-injured brain.
Cell reports (2023-04-13)
Ricardo L Azevedo-Pereira, Nathan C Manley, Chen Dong, Yue Zhang, Alex G Lee, Yulia Zatulovskaia, Varun Gupta, Jennifer Vu, Summer Han, Jack E Berry, Tonya M Bliss, Gary K Steinberg
ABSTRACT
Stem cell therapy shows promise for multiple disorders; however, the molecular crosstalk between grafted cells and host tissue is largely unknown. Here, we take a step toward addressing this question. Using translating ribosome affinity purification (TRAP) with sequencing tools, we simultaneously decode the transcriptomes of graft and host for human neural stem cells (hNSCs) transplanted into the stroke-injured rat brain. Employing pathway analysis tools, we investigate the interactions between the two transcriptomes to predict molecular pathways linking host and graft genes; as proof of concept, we predict host-secreted factors that signal to the graft and the downstream molecular cascades they trigger in the graft. We identify a potential host-graft crosstalk pathway where BMP6 from the stroke-injured brain induces graft secretion of noggin, a known brain repair factor. Decoding the molecular interplay between graft and host is a critical step toward deciphering the molecular mechanisms of stem cell action.
Sigma-Aldrich
Anti-phospho Smad1/Smad5/Smad8 (Ser463/465), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glutamate Transporter Antibody, Glial, serum, Chemicon®
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse