Merck
  • 18 bp insertion/duplication with internal missense mutation in human hepatic lipase gene exon 3. Mutations in brief no. 181. Online.
18 bp insertion/duplication with internal missense mutation in human hepatic lipase gene exon 3. Mutations in brief no. 181. Online.
Human mutation (2000-02-05)
O Tiebel, S Gehrisch, J Pietzsch, S Gromeier, W Jaross
ABSTRACT
Human hepatic lipase (hHL) plays an important role in hydrolysis of triglycerides from plasma lipoproteins. The enzyme also hydrolyzes HDL2 lipids resulting in smaller HDL particles with a lower cholesterol content and properties similar to HDL3. hHL is localized in liver sinusoids, ovary and adrenal gland. These findings propose an influence on processing of cholesterol. Here we report an insertion mutation in exon 3 of hHL. The 18 bp duplication contains an additional internal point mutation (GenBank-Accession #AF037404). The female mutation carrier suffered from severe adiposity with total cholesterol of 291,6mg/dl, HDL-cholesterol of 55,3mg/dl, LDL-cholesterol of 206,8mg/dl and triglycerides of 80,8mg/dl. Following cloning of a PCR-amplified fragment the mutation was confirmed by cycle sequencing. Sequence analysis revealed an inserted repeat of 18 nucleotides. Furthermore the patient carries an additional missense mutation A-->G at nucleotide 9 of the repeat which results in an amino acid exchange from Ile-->Val at codon 4 of the repeat. These data enable us to report the insertion of HisTyrThrValArgVal which might be responsible for the moderate shift in lipid metabolism of the heterozygous patient.
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)