Merck
  • Adiponutrin: A new gene regulated by energy balance in human adipose tissue.
Adiponutrin: A new gene regulated by energy balance in human adipose tissue.
The Journal of clinical endocrinology and metabolism (2004-06-08)
Yong-Ming Liu, Marthe Moldes, Jean-Philippe Bastard, Eric Bruckert, Nathalie Viguerie, Bernard Hainque, Arnaud Basdevant, Dominique Langin, Jacques Pairault, Karine Clément
ABSTRACT
Adiponutrin is a newly identified nonsecreted adipocyte protein regulated by changes in energy balance in rodents. We documented the influence of energy balance modification on adiponutrin gene expression in humans. We investigated the mRNA expression in sc adipose tissue of nonobese women and in obese women during 2-d very low-calorie diet (VLCD) and subsequent refeeding as well as before and after a VLCD of 3 wk (21-d VLCD). The adiponutrin mRNA levels of the nonobese and obese women were not different (P > 0.05). Two-day VLCD reduced the average level of adiponutrin mRNA expression by 36% (P = 0.0016), whereas refeeding elevated the mRNA level by 31% (P = 0.004). The 3-wk VLCD caused a dramatic 58% fall of the adiponutrin mRNA expression level (P = 0.001). The mRNA level was negatively correlated with fasting glucose (Rho = -0.62; P < 0.0001), and subjects with high adiponutrin mRNA level had an increased insulin sensitivity. Compared with other adipocyte proteins such as leptin and adiponectin, adiponutrin mRNA did not show correlation with either adiposity indexes or with leptin or adiponectin mRNAs. These results indicate that adiponutrin gene expression in humans is highly regulated by changes in energy balance.
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg