Merck
  • Novel PNLIPRP3 and DOCK8 gene expression and prognostic implications of DNA loss on chromosome 10q25.3 in hepatocellular carcinoma.
Novel PNLIPRP3 and DOCK8 gene expression and prognostic implications of DNA loss on chromosome 10q25.3 in hepatocellular carcinoma.
Asian Pacific journal of cancer prevention : APJCP (2009-07-31)
Pensri Saelee, Sopit Wongkham, Anucha Puapairoj, Narong Khuntikeo, Songsak Petmitr, Sunanta Chariyalertsak, Wutthi Sumethchotimaytha, Anant Karalak
ABSTRACT
Our previous study of gene alterations in 29 hepatocellular carcinoma (HCC) using AP-PCR amplified with 59 different 10-mer arbitrary primers and gene cloning, indicated DNA alterations by DNA fingerprints from 34 primers. Among these, the altered DNA fragment from primer U-8 predominated (62%). The aim of this report is to identify the gene alterations on chromosomal banding and gene expression in these patients, including the association of these alterations with patient demographic data. Seven different sequences, mapped to chromosomes 5q33.3, 7q31.33, 7q34, 9p24.3, 10q25.3, 13q31.3, and 16p11.2, were identified by gene cloning and nucleotide sequencing. Novel PNLIPRP3 gene over-expression and DOCK8 gene under-expression were observed in 41% and 44% of these patients, respectively, which point to an association of these genes and the development of HCC. Likewise, allelic loss on chromosome 10q25.3 was associated with shorter survival among HCC patients (P=0.03); this indicated that allelic loss on chromosome 10q25.3 may serve as a prognostic marker in patients with HCC.
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)