Merck
  • Human hepatic lipase. Cloned cDNA sequence, restriction fragment length polymorphisms, chromosomal localization, and evolutionary relationships with lipoprotein lipase and pancreatic lipase.
Human hepatic lipase. Cloned cDNA sequence, restriction fragment length polymorphisms, chromosomal localization, and evolutionary relationships with lipoprotein lipase and pancreatic lipase.
The Journal of biological chemistry (1988-01-25)
S Datta, C C Luo, W H Li, P VanTuinen, D H Ledbetter, M A Brown, S H Chen, S W Liu, L Chan
ABSTRACT
Human hepatic lipase is an important enzyme in high density lipoprotein (HDL) metabolism, being implicated in the conversion of HDL2 to HDL3. Three human hepatic lipase cDNA clones were identified in two lambda gt11 libraries from human liver. The cDNA-derived amino acid sequence predicts a protein of 476 amino acid residues, preceded by a 23-residue signal peptide. Four potential N-glycosylation sites are identified, two of which are conserved in rat hepatic lipase. On alignment with human, mouse, and bovine lipoprotein lipase, the same two sites were also conserved in lipoprotein lipase in all three species. Stringent conservation of the cysteine residues was also evident. Comparative analysis of amino acid sequences shows that hepatic lipase evolves at a rapid rate, 2.07 x 10(-9) substitutions/site/year, about four times that in lipoprotein lipase and half that in pancreatic lipase. Further, hepatic lipase and pancreatic lipase appear to be evolutionarily closer to each other than either of them is to lipoprotein lipase. Southern blot analysis revealed high frequency restriction fragment length polymorphisms of the hepatic lipase gene for the enzymes HindIII and MspI. these polymorphisms will be useful for haplotype and linkage analysis of the hepatic lipase gene. Using cloned human hepatic lipase cDNA as a hybridization probe, we performed Southern blot analysis of a panel of 13 human-rodent somatic cell hybrids. Concordance analysis of the various hybrid clones indicates that the hepatic lipase gene is located on the long arm of human chromosome 15. Analysis of hybrids containing different translocations of chromosome 15 localized the gene to the region 15q15----q22.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg