Merck
  • Plasma levels of pipecolic acid, both L- and D-enantiomers, in patients with chronic liver diseases, especially hepatic encephalopathy.
Plasma levels of pipecolic acid, both L- and D-enantiomers, in patients with chronic liver diseases, especially hepatic encephalopathy.
Clinica chimica acta; international journal of clinical chemistry (1999-10-06)
T Fujita, Y Amuro, T Hada, K Higashino
ABSTRACT
Pipecolic acid is regarded as a gamma-aminobutyric acid receptor agonist. Stereochemical studies of pipecolic acid were performed in patients with chronic liver diseases. Plasma D- and L-pipecolic acid were significantly elevated in 15 liver cirrhotic patients with no history of hepatic encephalopathy (1.05+/-0.24 micromol/l, 1.58+/-0.13 micromol/l, p < 0.01) and in 27 patients with chronic hepatic encephalopathy (1.58+/-0.50 micromol/l, 2.38+/-0.58 micromol/l, p<0.001) compared to 15 normal subjects. In seven patients with chronic hepatic encephalopathy orally treated with kanamycin, plasma pipecolic acid significantly decreased (D-acid: before 1.62+/-0.23 micromol/l, after 0.61+/-0.15 micromol/l; p<0.01, L-acid: before 2.43-0.52 micromol/l, after 2.23+/-0.11 micromol/l; p< 0.05). These results suggest that plasma pipecolic acid, particularly D-acid, is produced from D-lysine by intestinal bacteria in liver cirrhotic patients and that pipecolic acid could be involved in the pathogenesis of hepatic encephalopathy.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
Isovaleraldehyde, 97%
Sigma-Aldrich
Isovaleraldehyde, ≥97%, FG
Sigma-Aldrich
Isovaleraldehyde, natural, ≥95%, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
2-Heptanone, 99%