Merck
  • Dietary grape powder increases IL-1β and IL-6 production by lipopolysaccharide-activated monocytes and reduces plasma concentrations of large LDL and large LDL-cholesterol particles in obese humans.
Dietary grape powder increases IL-1β and IL-6 production by lipopolysaccharide-activated monocytes and reduces plasma concentrations of large LDL and large LDL-cholesterol particles in obese humans.
The British journal of nutrition (2014-05-17)
Susan J Zunino, Jan M Peerson, Tammy L Freytag, Andrew P Breksa, Ellen L Bonnel, Leslie R Woodhouse, David H Storms
ABSTRACT
Obese individuals are at an increased risk of developing CVD, hypertension, type 2 diabetes, and bacterial and viral infections when compared with the normal-weight population. In a 9-week randomised, double-blind, cross-over study, twenty-four obese subjects aged between 20 and 60 years and with a BMI between 30 and 45 kg/m2 were fed grape or placebo powder for 3-week intervals to determine the effects of dietary grapes on blood lipid profiles, plasma inflammatory marker concentrations and immune cell function. Blood samples were collected on days 1 and 8 for obtaining baseline information and at weeks 3, 4, 8 and 9. Comprehensive chemistry panels, lipid profile analyses by NMR, measurement of plasma inflammatory marker concentrations, and analyses of cytokine production by activated T lymphocytes and monocytes were performed for each blood draw. Dietary grape powder reduced the plasma concentrations of large LDL-cholesterol and large LDL particles compared with the placebo powder (P< 0·05). The concentrations of interferon-γ, TNF-α, IL-4 and IL-10 were measured in supernatants from peripheral blood mononuclear cells (PBMC) activated with anti-CD3/CD28 antibodies and those of TNF-α, IL-1β, IL-6 and IL-8 were measured in supernatants from PBMC activated with lipopolysaccharide (LPS). No difference in the production of T-cell cytokines was observed between the two intervention groups. The production of IL-1β and IL-6 was increased in supernatants from LPS-activated PBMC in the grape powder group compared with the placebo powder group (P< 0·05). These data suggest that dietary grapes may decrease atherogenic lipid fractions in obese individuals and increase the sensitivity of monocytes in a population at a greater risk of developing infections.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Fluorescein sodium salt, p.a.
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ultrapure Acetonitrile