Merck
  • Protective effect of olive oil and colocynth oil against cadmium-induced oxidative stress in the liver of Wistar rats.
Protective effect of olive oil and colocynth oil against cadmium-induced oxidative stress in the liver of Wistar rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-01-27)
Fouzia Amamou, Saïd Nemmiche, Radjaa Kaouthar Meziane, Amal Didi, Sidi Mohamed Yazit, Daoudi Chabane-Sari
ABSTRACT
Cadmium (Cd) is one of the most common heavy metal pollutants. It is accumulated particularly in liver and kidney. The present study examined the possible protective effect of olive oil and colocynth oil consumption against Cd-induced damage on plasma lipids and stress biochemical parameters of rats. Male albino Wistar rats were randomly divided into 6 groups of 5 animals each and treated orally with Cd (50 mg/l), olive oil and colocynth oil (4%) alone or in combination with cadmium for 8 weeks. It was shown that Cd exposure induced significant increases in the activities of serum alanine aminotransferase, aspartate aminotransferase, lipid peroxidation levels (MDA) and protein carbonyl contents in exposed groups of rats compared to control group while the antioxidant enzymes, reduced glutathione and vitamins (C, A and E) were significantly decreased. Co-treatment with olive oil or colocynth oil significantly improved the oxidative damage induced by Cd. The antioxidant potential in plasma and liver were markedly restored with a significant decline in MDA levels and activity of transaminases. In conclusion, these results suggest that olive oil or colocynth oil consumption could protect the rat liver against Cd-induced injury by increasing the activities of antioxidant enzymes and reducing oxidative stress.
Product Description
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Water, for TOC analysis
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Methanol, analytical standard
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)