Merck
  • Changes in carotenoid profiles and in the expression pattern of the genes in carotenoid metabolisms during fruit development and ripening in four watermelon cultivars.
Changes in carotenoid profiles and in the expression pattern of the genes in carotenoid metabolisms during fruit development and ripening in four watermelon cultivars.
Food chemistry (2014-12-23)
Pin Lv, Na Li, Hui Liu, Huihui Gu, Wen-En Zhao
ABSTRACT
Changes in carotenoid profiles during fruit ripening were investigated in four watermelon cultivars: red-fleshed "CN66", pink-fleshed "CN62", yellow-fleshed "ZXG381" and white-fleshed "ZXG507". The expression pattern of twelve genes (GGPS, PSY, PSY-A, PDS, ZDS, CRTISO, LCYB, CHYB, ZEP, NCED1, NCED2 and NCED3) was analysed. In "CN66" and "CN62", lycopene appeared at 12 DAP and became a main carotenoid increased at the later stages. The transcript levels of carotenogenic genes in "CN66" sharply increased during 18-30 DAP, and concomitantly, fruit accumulated the massive amounts of carotenoids. In "ZXG381", violaxanthin and lutein contents were positively correlated, respectively, with CHYB and ZEP transcript levels during fruit ripening. The trace amounts of carotenoids in "ZXG507" were accompanied with the low transcript levels of most biosynthetic genes. The results suggest that differential transcriptional regulation of carotenoid metabolic genes is very important in determining the amount and type of specific carotenoids accumulated during fruit development and ripening.
Sigma-Aldrich
Lycopene, Redivivo, 10% FS, ~10% in corn oil, ≥95.0% (sum of isomers)
Supelco
Lycopene, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Acetonitrile, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Lutein, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%