Merck
  • Norisoboldine attenuates inflammatory pain via the adenosine A1 receptor.
Norisoboldine attenuates inflammatory pain via the adenosine A1 receptor.
European journal of pain (London, England) (2014-01-08)
X Gao, Q Lu, G Chou, Z Wang, R Pan, Y Xia, H Hu, Y Dai
ABSTRACT
Norisoboldine (NOR) is a benzylisoquinoline alkaloid isolated from Radix Linderae, a traditional Chinese medicine. Our previous studies have demonstrated that it produces anti-inflammatory and anti-rheumatoid arthritis effects. The present study was undertaken to explore the analgesic effects of NOR and its potential mechanism in the formalin test and the acetic acid writhing test. Oral administration of NOR dose dependently attenuated the formalin-induced pain responses in the second phase, and reduced formalin-induced paw oedema. It also diminished acetic acid-induced writhing responses but had no effect on acute thermal pain in the hotplate test. The mechanistic studies suggested that the adenosine system, but not the opioid receptor system, is involved in NOR-induced antinociception. Naloxone, a non-selective opioid receptor antagonist, had no effect on NOR-induced analgesic action. However, caffeine (a non-selective adenosine receptor antagonist) completely reversed the analgesic effect of NOR in formalin-induced nociceptive responses in the second phase, and 8-cyclopentyl-1, 3-dipropylxanthine (DPCPX, a selective adenosine A1 receptor antagonist) completely inhibited NOR-induced analgesia in both formalin-induced nociceptive responses and acetic acid-induced writhing responses. In addition, NOR reduced formalin-induced activation of extracellular signal-regulated kinase and calcium/calmodulin-dependent protein kinase II in the spinal cord, which is also blocked by DPCPX. Furthermore, NOR decreased forskolin-evoked cyclic adenosine monophosphate levels in mouse spinal cord neuronal cultures through the adenosine A1 receptor. Our data demonstrate that NOR produces the analgesic effect in inflammatory pain by a mechanism related to the adenosine system.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
2-Chloro-N6-cyclopentyladenosine, adenosine receptor agonist
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Forskolin, analytical standard
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.