Merck
  • Myosin VIIA regulates microvillus morphogenesis and interacts with cadherin Cad99C in Drosophila oogenesis.
Myosin VIIA regulates microvillus morphogenesis and interacts with cadherin Cad99C in Drosophila oogenesis.
Journal of cell science (2014-09-23)
Cory Glowinski, Ri-Hua Sandy Liu, Xi Chen, Audrey Darabie, Dorothea Godt
ABSTRACT
Microvilli and related actin-based protrusions permit multiple interactions between cells and their environment. How the shape, length and arrangement of microvilli are determined remains largely unclear. To address this issue and explore the cooperation of the two main components of a microvillus, the central F-actin bundle and the enveloping plasma membrane, we investigated the expression and function of Myosin VIIA (Myo7A), which is encoded by crinkled (ck), and its interaction with cadherin Cad99C in the microvilli of the Drosophila follicular epithelium. Myo7A is present in the microvilli and terminal web of follicle cells, and associates with several other F-actin-rich structures in the ovary. Loss of Myo7A caused brush border defects and a reduction in the amount of the microvillus regulator Cad99C. We show that Myo7A and Cad99C form a molecular complex and that the cytoplasmic tail of Cad99C recruits Myo7A to microvilli. Our data indicate that Myo7A regulates the structure and spacing of microvilli, and interacts with Cad99C in vivo. A comparison of the mutant phenotypes suggests that Myo7A and Cad99C have co-dependent and independent functions in microvilli.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Glutaraldehyde solution, 25 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
(±)-Propylene oxide, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard