Merck
  • Rational design of esterase BioH with enhanced enantioselectivity towards methyl (S)-o-chloromandelate.
Rational design of esterase BioH with enhanced enantioselectivity towards methyl (S)-o-chloromandelate.
Applied microbiology and biotechnology (2014-08-12)
Jiali Gu, Lidan Ye, Fei Guo, Xiaomei Lv, Wenqiang Lu, Hongwei Yu
ABSTRACT
Methyl (R)-o-chloromandelate (R-CMM) is an intermediate for the platelet aggregation inhibitor clopidogrel. Its preparation through enzymatic resolution of the corresponding ester has been hindered by the lack of an enzyme with satisfying enantioselectivity and activity. In the present work, we aimed to improve the enzymatic enantioselectivity towards methyl (S)-o-chloromandelate (S-CMM) by rational design, using esterase BioH as a model enzyme. Based on the differences in the binding mode of S- and R-enantiomers at the active cavity of the enzyme, the steric and electronic interactions between the key amino acids of BioH and the enantiomers were finely tuned. The enantioselectivity of esterase BioH towards S-CMM was improved from 3.3 (the wild type) to 73.4 (L123V/L181A/L207F). Synergistic interaction was observed between point mutations, and insight into the source of enzymatic enantioselectivity was gained by molecular dynamics simulations. The results can provide a reference for the enzyme design of other enzymes towards S-CMM for the enhancement of enantioselectivity.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Ethyl acetate, analytical standard
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%