Merck
  • Ulcer healing and mechanism(s) of action involved in the gastroprotective activity of fractions obtained from Syngonanthus arthrotrichus and Syngonanthus bisulcatus.
Ulcer healing and mechanism(s) of action involved in the gastroprotective activity of fractions obtained from Syngonanthus arthrotrichus and Syngonanthus bisulcatus.
BMC complementary and alternative medicine (2015-10-30)
Leônia Maria Batista, Gedson Rodrigues De Morais Lima, Ana Beatriz Albino De Almeida, Luciana De Pietro Magri, Tamara Regina Calvo, Anderson Luiz Ferreira, Cláudia Helena Pellizzon, Clélia Akiko Hiruma-Lima, Wagner Vilegas, Paulo Takeo Sano, Alba Regina Monteiro Souza Brito
ABSTRACT
Syngonanthus arthrotrichus and Syngonanthus bisulcatus, currently known for Comanthera aciphylla (Bong.) L.R.Parra & Giul. and Comanthera bisulcata (Koern.) L.R. Parra & Giul, popularly known in Brazil as "sempre-vivas," are plants from the family Eriocaulaceae. They are found in the states of Minas Gerais and Bahia. The species are known to be rich in flavonoids to which their gastroprotective activity has been attributed. In this research, experimental protocols were performed to elucidate the associated mechanisms of action. The activity was evaluated using induced gastric ulcer models (acetic acid and ethanol-induced gastric lesions in NEM or L-NAME pre-treated mice, and by ischemia/reperfusion). Antioxidant enzymes, serum somatostatin, and gastrin were also evaluated. In chronic gastric ulcers, a single daily oral dose of Sa-FRF or Sb-FRF (100 mg/kg body wt.) for 14 consecutive days accelerated ulcer healing to an extent similar to that seen with an equal dose of cimetidine. The pre-treatment of mice with NEM (N-ethylmaleimide) or L-NAME (N-nitro-L-arginine) abolished the protective activity of Sa-FRF, Sa-FDF, Sb-FDF and Sb-FRF or Sa-FRF and Sb-FRF, respectively, which indicates that antioxidant compounds and nitric oxide synthase activity are involved in the gastroprotective. Sa-FRF and Sb-FRF (100 mg/kg p.o) protected the gastric mucosa against ulceration that was induced by ischemia/reperfusion (72 and 76 %, respectively). It also decreased lipid peroxidation and restored total thiols in the gastric wall of mice that had been treated with ethanol. When administered to rats submitted to ethanol-induced gastric lesions, Sa-FRF and Sb-FRF (100 mg/kg, p.o.) increased the somatostatin serum levels, while the gastrin serum levels were proportionally decreased. The results indicate significant healing effects and gastroprotective activity for the Sa-FRF and Sb-FRF, which probably involves the participation of SH groups, nitric oxide (NO), the antioxidant system, somatostatin, and gastrin. All are integral parts of the gastrointestinal mucosa's cytoprotective mechanisms against aggressive factors.
Product Description
Supelco
Chloromethane solution, 200 μg/mL in methanol, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Lansoprazole, ≥98% (TLC), powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
Chloromethane solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Chloromethane solution, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%