Merck
  • Uterine Leiomyosarcoma Tumorigenesis in Lmp2-deficient Mice: Involvement of Impaired Anti-oncogenic Factor IRF1.
Uterine Leiomyosarcoma Tumorigenesis in Lmp2-deficient Mice: Involvement of Impaired Anti-oncogenic Factor IRF1.
Anticancer research (2015-08-09)
Takuma Hayashi, Akiko Horiuchi, Kenji Sano, Nobuo Yaegashi, Ikuo Konishi
ABSTRACT
Uterine leiomyosarcoma (Ut-LMS) is a highly metastatic smooth muscle neoplasm. We have previously reported that low molecular mass protein2 Lmp2-deficient mice spontaneously developed Ut-LMS, which implicated this protein as an anti-oncogenic candidate. We also suggested that LMP2 may negatively regulate Ut-LMS independently of its role in the proteasome. Initially described as a transcription factor able to activate the expression of interferon-gamma (IFN-γ)-responsive genes, interferon regulatory factor-1 (IRF1) has been shown to play roles in the immune response, and tumor suppression. The aim of this study was to elucidate the molecular mechanism of sarcomagenesis of Ut-LMS using human and mouse uterine tissues. The expression of the IFN-γ signal molecules, IRF1 and -2, STAT1, and LMP2, -3, -7 and -10 were examined by western blot analysis, electrophoretic mobility shift assay and immunohistochemistry in human and mouse uterine tissues. Physiological significance of IRF1 in sarcomagenesis of Ut-LMS was demonstrated by xenograft studies. In the present study, several lines of evidence indicated that although treatment with IFN-γ strongly induced the activation of STAT1 as a transcriptional activator, its target molecule, IRF1, was not clearly produced in Lmp2-deficient uterine smooth muscle cells (Ut-SMCs). Defective expression of IRF1 in the IFN-γ-induced signaling molecules may result in the malignant transformation of Ut-SMCs. The modulation of LMP2 may lead to new therapeutic approaches in human Ut-LMS.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Tumor Necrosis Factor-α from mouse, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)