Merck
  • Mechanism of Polyphosphates Hydrolysis by Purified Polyphosphatases from the Dorsal Muscle of Silver Carp (Hypophthalmichthys Molitrix) as Detected by ³¹P NMR.
Mechanism of Polyphosphates Hydrolysis by Purified Polyphosphatases from the Dorsal Muscle of Silver Carp (Hypophthalmichthys Molitrix) as Detected by ³¹P NMR.
Journal of food science (2015-09-30)
Wei Liu, Meng Xu, Yawei Zhang, Fulong Wang, Teng Hui, Baowei Cui, Xiuyun Guo, Zengqi Peng
ABSTRACT
The dynamic hydrolysis of tetrasodium pyrophosphate (TSPP), sodium tripolyphosphate (STPP) and polyphosphate compound, which was catalyzed by purified pyrophosphatase (PPase) and myosin- tripolyphosphatase (TPPase) from the silver carp dorsal muscle, was studied using (31) P NMR spectroscopy. In the PPase + TSPP system, the pyrophosphate (PP) was hydrolyzed quickly and completely within 8 h and the hydrolysis rate of PP was 12.51%/h. In the TPPase + STPP system, the first-order hydrolysis of tripolyphosphate (TPP) was not yet complete after 48 h, and the derived PP accumulated progressively. Given the coexistence of PPase and TPPase, only 1.20% of TPP in STPP alone remained after 48 h. However, the generation rate of Pi in the polyphosphate compound (TSPP: STPP: sodium hexametaphosphate = 1: 8: 1) was 0.76%/h, which was less than 0.88%/h in STPP alone. In the presence of polyphosphatases, the decrease of PP or TPP content in the polyphosphate compound was not as rapid as that in TSPP or STPP alone due to the inhibitory effect of PP on TPPase and the effect of low system pH on PPase. The understanding of polyphosphates hydrolysis mechanism was capable of developing the advanced polyphosphate mixture in order to reduce the phosphate residue in fish products. Processors appreciate the proven value of phosphates to increase the yield and functionality of the fish meat products. Our studies showed that the hydrolysis rate of PP or TPP in the blend was slower than that of polyphosphate alone. Thus, it is likely that the addition of PP and TPP in a polyphosphate blend had a prolonged interaction with proteins in fish meat processing and the effectiveness of polyphosphates was enhanced.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D