Merck
SAMPHV001
Millex Samplicity® filter
PVDF membrane, pore size 0.45 μm, diam. 25 mm, non-sterile, hydrophilic
Synonym(s):
0.45 μm PVDF syringe filter, Durapore® PVDF syringe filter, Millex-HV syringe filter, disposable syringe filter, syringe filter
Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41104922
eCl@ss:
32031690
NACRES:
NB.24
Skip To
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PVDF membrane
high-density polyethylene
sterility
non-sterile
product line
Millex
feature
hydrophilic
manufacturer/tradename
Samplicity®
technique(s)
HPLC: suitable
diam.
25 mm
volume
1.7 mL
pore size
0.45 μm pore size
fitting
inlet funnel (Easy loading)
outlet tip (for smooth transfer to HPLC vials)
shipped in
ambient
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
SAMPLCR01SAMPLG001SAMP2HVNB
material
PVDF membrane, high-density polyethylene
material
PTFE membrane, high-density polyethylene
material
PTFE membrane, high-density polyethylene
material
PVDF membrane, polypropylene housing
pore size
0.45 μm pore size
pore size
0.45 μm pore size
pore size
0.2 μm pore size
pore size
0.45 μm
manufacturer/tradename
Samplicity®
manufacturer/tradename
Samplicity®
manufacturer/tradename
Samplicity®
manufacturer/tradename
Millex, Samplicity®
diam.
25 mm
diam.
25 mm
diam.
25 mm
diam.
33 mm
fitting
inlet funnel (Easy loading), outlet tip (for smooth transfer to HPLC vials)
fitting
inlet funnel (Easy loading), outlet tip (for smooth transfer to HPLC vials)
fitting
inlet funnel (Easy loading), outlet tip (for smooth transfer to HPLC vials)
fitting
female Luer-Lok® inlet connection, female Luer-Lok® inlet fitting, male Luer outlet slip
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
General description
SAMP2HVNB replaces SAMPHV001
Overview of Millex Samplicity® Filters for the Samplicity® G2 System
  • Samplicity® G2 Filtration System enables vacuum driven filtation of up to 8 samples at once, directly into HPLC vials
  • Filtration for HPLC sample prep without syringes
  • Millex Samplicity®Filters provide integrated funnels and filters for easiest sample loading
Application
To be used with the Samplicity Filtration System. Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLEX SAMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
Millex is a trademark of Merck KGaA, Darmstadt, Germany
SAMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Related Content
HPLC sample prep doesn’t have to be inefficient or painful. The ideal sample prep system for most chromatographers. If you’re using chromatography to analyze dozens of pharmaceutical samples, food and beverage products or cosmetic formulations a day, especially hard-to-filter, viscous or particulate-laden samples, you know that traditional filtration methods can result in: Slow filtration speed/clogging, Filter bursting or spillage, Sample volume loss (most undesirable for small-volume samples), and Operator fatigue. The first vacuum-driven system with the flexibility to filter 1 to 8 samples directly into standard HPLC sample vials, the Samplicity® Filtration System addresses all of the challenges facing analytical chromatographers. Just attach a vacuum pump, load your samples, and flip the lever. Recover your particulate-free samples in seconds.
See All
Questions
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service