Skip to Content
Merck

AABH14DCDC8E

AA BLOCKS, INC.

4'-Tert-butyl[1,1'-biphenyl]-2-carboxylic acid

Synonym(s):

4'-tert-butyl[1,1'-biphenyl]-2-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C17H18O2
Molecular Weight:
254.33
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C17H18O2/c1-17(2,3)13-10-8-12(9-11-13)14-6-4-5-7-15(14)16(18)19/h4-11H,1-3H3,(H,18,19)

Inchi Key

WYIJUURMGVBXEM-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library