Skip to Content
Merck

AABH30371CDA

AA BLOCKS, INC.

8-Methoxymethyl-3-isobutyl-1-methylxanthine

Synonym(s):

3-isobutyl-8-(methoxymethyl)-1-methyl-3,7-dihydro-1H-purine-2,6-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H18O3N4
CAS Number:
Molecular Weight:
266.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H18N4O3/c1-7(2)5-16-10-9(11(17)15(3)12(16)18)13-8(14-10)6-19-4/h7H,5-6H2,1-4H3,(H,13,14)

Inchi Key

NBLBCGUCPBXKOV-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Storage temp.

2~8C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library