Inchi Code
1S/C26H26N3O.ClHO4/c27-15-22-20-13-16-5-1-9-28-11-3-7-18(23(16)28)25(20)30-26-19-8-4-12-29-10-2-6-17(24(19)29)14-21(22)26;2-1(3,4)5/h13-14H,1-12H2;(H,2,3,4,5)/q+1;/p-1
Inchi Key
HACOCUMLBPNDIN-UHFFFAOYSA-M
Country of Origin
US
Purity
98%
Storage temp.
2~8C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.