Skip to Content
Merck

AABH93DE42E4

AA BLOCKS, INC.

DAF-2 diacetate

Synonym(s):

5,6-diamino-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H18O7N2
CAS Number:
Molecular Weight:
446.42
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C24H18N2O7/c1-11(27)30-13-3-5-16-21(7-13)32-22-8-14(31-12(2)28)4-6-17(22)24(16)18-10-20(26)19(25)9-15(18)23(29)33-24/h3-10H,25-26H2,1-2H3

Inchi Key

PTSUYDXEEKDBQU-UHFFFAOYSA-N

Storage temp.

2-8C

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library