Skip to Content
Merck

AABH97CD5FB3

AA BLOCKS, INC.

Erythromycin Lactobionate

Synonym(s):

(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-((2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yloxy)-14-ethyl-7,12,13-trihydroxy-4-((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yloxy)-3,5,7,9,11,13-hexamethyloxacyclotetradecane-2,10-dione (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yloxy)hexanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Storage temp.

2~8C

Inchi Code

1S/C37H67NO13.C12H22O12/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26;13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3;3-10,12-20H,1-2H2,(H,21,22)/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-;3-,4-,5+,6+,7-,8-,9-,10-,12+/m11/s1

Inchi Key

NNRXCKZMQLFUPL-WBMZRJHASA-N

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library