Skip to Content
Merck

AABH97CD6A71

AA BLOCKS, INC.

(S)-2-Amino-N-(4-nitro-phenyl)-propionamide hydrochloride

Synonym(s):

(2S)-2-amino-N-(4-nitrophenyl)propanamide hydrochloride

Select a Size

Change View

About This Item

Linear Formula:
C9H12O3N3Cl1
CAS Number:
Molecular Weight:
245.67
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11N3O3.ClH/c1-6(10)9(13)11-7-2-4-8(5-3-7)12(14)15;/h2-6H,10H2,1H3,(H,11,13);1H/t6-;/m0./s1

Inchi Key

YEXRLSXNWLNHQR-RGMNGODLSA-N

Purity

99%

Storage temp.

RT

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library