Skip to Content
Merck

AABH97CD71F3

AA BLOCKS, INC.

Fangchinoline

Synonym(s):

(1beta)-6,6',12-trimethoxy-2,2'-dimethylberbaman-7-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C37H40O6N2
CAS Number:
Molecular Weight:
608.73
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C37H40N2O6/c1-38-14-12-24-19-31(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-32-18-23(8-11-30(32)41-3)17-29-35-25(13-15-39(29)2)20-34(43-5)36(40)37(35)45-33/h6-11,18-21,28-29,40H,12-17H2,1-5H3/t28-,29-/m0/s1

Inchi Key

IIQSJHUEZBTSAT-VMPREFPWSA-N

Purity

98%

Storage temp.

2-8C

Country of Origin

US



Still not finding the right product?

Explore all of our products under Fangchinoline




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library